Proteintech
Manufacturer Code:179961AP150UL
Catalog # 50-173-1925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | APOA4 Fusion Protein Ag12454 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apo-AIV; apoA-IV; APOA4 apolipoprotein A IV; Apolipoprotein A-IV; Apolipoprotein A4
Gene Aliases: Apo-AIV; Apoa-4; ApoA-IV; APOA4; apoAIV
UniProt ID: (Human) P06727, (Mouse) P06728, (Rat) P02651
Entrez Gene ID: (Human) 337, (Mouse) 11808, (Rat) 25080
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.