Invitrogen
Manufacturer Code:PA518975
Catalog # PIPA518975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PPYPDTECVYRLQ) corresponding to the C-terminus amino acids of APOA1BP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AI-BP; apoA-I binding protein; APOA1BP apolipoprotein A-I binding protein AIBP MGC119143 MGC119144 MGC119145 apoA-I binding protein; apolipoprotein A-I binding protein; Apolipoprotein A-I-binding protein; NAD(P)H-hydrate epimerase; NAD(P)HX epimerase; YjeF N-terminal domain-containing protein 1; YjeF_N1
Gene Aliases: AIBP; APOA1BP; NAXE; YJEFN1
UniProt ID: (Human) Q8NCW5
Entrez Gene ID: (Human) 128240
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.