Invitrogen
Manufacturer Code:PA5116647
Catalog # PIPA5116647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 48-170 of human NAXE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA087124 Aibp AI-BP AI-BP {ECO:0000255 APOA1BP Apoa1ip apoA-I binding protein apolipoprotein A-I binding protein apolipoprotein A-I interacting protein apolipoprotein A-I-binding protein apolipoprotein A-I-binding protein {ECO:0000255 ESTM37 HAMAP-Rule:MF_03159} MGC119143 MGC119144 MGC119145 NAD(P)H-hydrate epimerase NAD(P)H-hydrate epimerase {ECO:0000255 NAD(P)HX epimerase NAD(P)HX epimerase {ECO:0000255 Naxe yjeF N-terminal domain-containing protein 1 YjeF_N1 YJEFN1; AI-BP; AI-BP {ECO:0000255|HAMAP-Rule:MF_03159}; apoA-I binding protein; apolipoprotein A-I binding protein; apolipoprotein A-I interacting protein; Apolipoprotein A-I-binding protein; apolipoprotein A-I-binding protein {ECO:0000255|HAMAP-Rule:MF_03159}; NAD(P)H-hydrate epimerase; NAD(P)H-hydrate epimerase {ECO:0000255|HAMAP-Rule:MF_03159}; NAD(P)HX epimerase; NAD(P)HX epimerase {ECO:0000255|HAMAP-Rule:MF_03159}; YjeF N-terminal domain-containing protein 1; YjeF_N1
Gene Aliases: AA087124; AI-BP; AIBP; APOA1BP; Apoa1ip; ESTM37; NAXE; YJEFN1
UniProt ID: (Rat) B0BNM1, (Mouse) Q8K4Z3, (Human) Q8NCW5
Entrez Gene ID: (Rat) 295229, (Mouse) 246703, (Human) 128240
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.