Novus Biologicals
Manufacturer Code:NBP292287100UL
Catalog # NB026465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 48-170 of human NAXE (NP_658985.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI-BP; AIBPAI-BP apolipoprotein A-I binding protein apolipoprotein A-I-binding protein MGC119143 MGC119144 MGC119145 YjeF N-terminal domain-containing protein 1 YjeF_N1 YJEFN1apoA-I binding protein; apoA-I binding protein; apolipoprotein A-I binding protein; Apolipoprotein A-I-binding protein; NAD(P)H-hydrate epimerase; NAD(P)HX epimerase; YjeF N-terminal domain-containing protein 1; YjeF_N1
Gene Aliases: AIBP; APOA1BP; NAXE; YJEFN1
UniProt ID: (Human) Q8NCW5
Entrez Gene ID: (Human) 128240
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.