Novus Biologicals
Manufacturer Code:NBP230736
Catalog # NBP230736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LKQQVEGTAQEAVSAAGAAAQQVVDQATEAGQKAMDQLAKTTQETID |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adipogenesis factor rich in obesity; Adipogenesis regulatory factor; Adipose most abundant gene transcript 2 protein; adipose specific 2; adipose specific 2 apM-2 APM2 chromosome 10 open reading frame 116; Adipose-specific protein 2; apM-2
Gene Aliases: ADIRF; AFRO; apM-2; APM2; C10orf116
UniProt ID: (Human) Q15847
Entrez Gene ID: (Human) 10974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.