Invitrogen
Manufacturer Code:PA578795
Catalog # PIPA578795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human APLP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amyloid beta (A4) precursor-like protein 1; amyloid precursor-like protein 1; Amyloid-like protein 1; Amyloid-like protein 1 APLP APLP-1 C30 APLP1; APLP; APLP-1; C30
Gene Aliases: APLP; APLP1; Aplp1_retired; RGD1561211
UniProt ID: (Human) P51693, (Mouse) Q03157
Entrez Gene ID: (Human) 333, (Rat) 502317, (Mouse) 11803
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.