Invitrogen
Manufacturer Code:PA577051
Catalog # PIPA577051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human APLP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amyloid beta (A4) precursor-like protein 1; amyloid precursor-like protein 1; Amyloid-like protein 1; Amyloid-like protein 1 precursorAPLP 1APLPAPLP-1Aplp1APLP1&lowbarHUMANC30; APLP; APLP-1; C30
Gene Aliases: APLP; APLP1; Aplp1_retired; RGD1561211
UniProt ID: (Human) P51693, (Mouse) Q03157
Entrez Gene ID: (Human) 333, (Rat) 502317, (Mouse) 11803
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.