Invitrogen
Manufacturer Code:PA5109399
Catalog # PIPA5109399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 39-300 of human APLP1 (NP_005157.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amyloid beta (A4) precursor-like protein 1; amyloid precursor-like protein 1; Amyloid-like protein 1; Amyloid-like protein 1 APLP APLP 1 Aplp1 Aplp1_retired APLPAPLP-1 C30 RGD1561211; APLP; APLP-1; C30
Gene Aliases: APLP; APLP1; Aplp1_retired; RGD1561211
UniProt ID: (Human) P51693, (Mouse) Q03157
Entrez Gene ID: (Human) 333, (Rat) 502317, (Mouse) 11803
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.