Invitrogen
Manufacturer Code:PA572239
Catalog # PIPA572239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 343-377 amino acids from the human region of DANRE aplnra |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiotensin II receptor-like 1a; angiotensin receptor-like 1; Angiotensin receptor-like 1a; Apelin receptor A; Apelin receptor A Angiotensin II receptor-like 1a Angiotensin receptor-like 1a G-protein coupled receptor APJ A aplnra agtrl1 {ECO:0000312; EMBL:ABI994701} agtrl1a; G-protein coupled receptor APJ A
Gene Aliases: Agtr1; agtrl1; agtrl1a; aplnra; msr; zgc:152843
UniProt ID: (Zebrafish) Q7SZP9
Entrez Gene ID: (Zebrafish) 561935
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.