Abnova
Manufacturer Code:MAB0105
Catalog # 89-107-873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 19F461 |
Immunogen | Recombinant His fusion protein corresponding to full length human APIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant APIP.
For Research Use Only
Protein Aliases: APAF1-interacting protein; APIP; APIP2; CGI-29; CGI29; dJ179L10.2; hAPIP; Methylthioribulose-1-phosphate dehydratase; MMRP19; MTRu-1-P dehydratase; probable methylthioribulose-1-phosphate dehydratase
Gene Aliases: APIP; APIP2; CGI-29; CGI29; hAPIP; MMRP19
UniProt ID: (Human) Q96GX9
Entrez Gene ID: (Human) 51074
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.