Invitrogen
Manufacturer Code:PA577133
Catalog # PIPA577133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human APIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APAF1-interacting protein; APIP; hAPIP; Methylthioribulose-1-phosphate dehydratase; Methylthioribulose-1-phosphate dehydrataseUniRule annotation MTRu-1-P dehydrataseUniRule annotation APAF1-interacting proteinUniRule annotation hAPIP APIP CGI-29; monocyte macrophage 19; Monocyte/macrophage protein 19; MTRu-1-P dehydratase; probable methylthioribulose-1-phosphate dehydratase
Gene Aliases: APIP; APIP2; CGI-29; CGI29; hAPIP; MMRP19; RGD1564562
UniProt ID: (Human) Q96GX9, (Mouse) Q9WVQ5
Entrez Gene ID: (Human) 51074, (Rat) 295961, (Mouse) 56369
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.