Proteintech
Manufacturer Code:120892AP150UL
Catalog # 50-172-7704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | APIP Fusion Protein Ag2729 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APAF1 interacting protein APIP APIP2 CGI 29 CGI29 MMRP19 MTRu 1 P dehydratase; APAF1-interacting protein; APIP; hAPIP; Methylthioribulose-1-phosphate dehydratase; monocyte macrophage 19; Monocyte/macrophage protein 19; MTRu-1-P dehydratase; probable methylthioribulose-1-phosphate dehydratase
Gene Aliases: APIP; APIP2; CGI-29; CGI29; hAPIP; MMRP19; RGD1564562
UniProt ID: (Human) Q96GX9, (Mouse) Q9WVQ5
Entrez Gene ID: (Human) 51074, (Rat) 295961, (Mouse) 56369
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.