Novus Biologicals
Manufacturer Code:NB600471
Catalog # NB600471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This purified antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast APG8 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: APG8 Atg8p AUT7 CVT5; Autophagy-related protein 8; Autophagy-related ubiquitin-like modifier ATG8; Cytoplasm to vacuole targeting protein 5; ubiquitin-like protein ATG8
Gene Aliases: APG8; ATG8; AUT7; CVT5; YBL0732; YBL078C
UniProt ID: (Baker's yeast) P38182
Entrez Gene ID: (Baker's yeast) 852200
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.