Novus Biologicals
Manufacturer Code:NB100116SS
Catalog # NB100116SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Gel Shift (GS) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13B8E5C2 |
Immunogen | Purified human APE1 [Uniprot: P27695] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AP endonuclease 1; AP endonuclease 1 APE APE1 APE-1 APEAPEX nuclease (multifunctional DNA repair enzyme) APENAP endonuclease class I APEX deoxyribonuclease (apurinic or apyrimidinic) APEX nuclease APEX nuclease (multifunctional DNA repair enzyme) 1 Apurinic-apyrimidinic endonuclease 1 APXAP lyase DNA-(apurinic or apyrimidinic site) lyase EC 3.1 EC 4.2.99.18 HAP1apurinic/apyrimidinic (abasic) endonuclease multifunctional DNA repair enzyme protein REF-1 redox factor 1 Redox factor-1 REF-1 REF1 apurinic/apyrimidinic exonuclease; AP endonuclease class I; AP lyase; APEN; APEX nuclease; APEX nuclease (multifunctional DNA repair enzyme) 1; Apurinic-apyrimidinic endonuclease 1; apurinic/apyrimidinic (abasic) endonuclease; deoxyribonuclease (apurinic or apyrimidinic); DNA-(apurinic or apyrimidinic site) endonuclease; DNA-(apurinic or apyrimidinic site) endonuclease, mitochondrial; protein REF-1; Redox factor-1; REF-1
Gene Aliases: APE; APE1; APEN; APEX; APEX1; APX; HAP1; REF1
UniProt ID: (Human) P27695
Entrez Gene ID: (Human) 328
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.