OriGene
Manufacturer Code:TA505247
Catalog # 50-167-5767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D6 |
Immunogen | Full length human recombit protein of human APCS produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 9.5S alpha-1-glycoprotein; epididymis secretory sperm binding protein Li 92n; HEL-S-92n PTX2 SAP; pentaxin-related; pentraxin-2; pentraxin-related; SAP; Serum amyloid P-component; Serum amyloid P-component(1-203)
Gene Aliases: APCS; HEL-S-92n; PTX2; SAP
UniProt ID: (Human) P02743
Entrez Gene ID: (Human) 325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.