Abnova
Manufacturer Code:H00000325M11
Catalog # 89-016-165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F3 |
Immunogen | APCS (AAH07058.1 35 a.a. ~ 149 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant APCS.
For Research Use Only
Protein Aliases: 9.5S alpha-1-glycoprotein; epididymis secretory sperm binding protein Li 92n; MGC88159; pentaxin-related; pentraxin-2; pentraxin-related; PTX2; SAP; Serum amyloid P-component; Serum amyloid P-component(1-203)
Gene Aliases: APCS; HEL-S-92n; PTX2; SAP
UniProt ID: (Human) P02743
Entrez Gene ID: (Human) 325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.