Abnova
Manufacturer Code:PAB29711
Catalog # 89-933-571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified rat Apcs. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against rat Apcs.
For Research Use Only
Protein Aliases: SAP; Serum amyloid P-component
Gene Aliases: Apcs; Ptx2; Sap
UniProt ID: (Rat) P23680
Entrez Gene ID: (Rat) 29339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.