Invitrogen
Manufacturer Code:MA526262
Catalog # PIMA526262
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F2 |
Immunogen | Full length human recombit protein of ANAPC11 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: anaphase promoting complex subunit 11 (yeast APC11 homolog); Anaphase-promoting complex subunit 11; APC11; APC11 anaphase promoting complex subunit 11 homolog; Cyclosome subunit 11; Hepatocellular carcinoma-associated RING finger protein; HSPC214 APC11 Apc11p APC11 anaphase promoting complex subunit 11 homolog anaphase promoting complex subunit 11 (yeast APC11 homolog) anaphase-promoting complex subunit 11 cyclosome subunit 11 hepatocellular carcinoma-associated RING finger protein
Gene Aliases: ANAPC11; APC11; Apc11p; HSPC214
UniProt ID: (Human) B7ZW64
Entrez Gene ID: (Human) 51529
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.