Proteintech
Manufacturer Code:197821AP150UL
Catalog # 50-173-2538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli APC BTPS2 Deleted in polyposis 2.5 DP2 DP2.5 DP3 GS Protein APC; Adenomatous polyposis coli protein; Deleted in polyposis 2.5; multiple intestinal neoplasia; Protein APC; protein phosphatase 1, regulatory subunit 46; truncated adenomatosis polyposis coli; WNT signaling pathway regulator
Gene Aliases: AI047805; APC; AU020952; AW124434; BTPS2; CC1; DP2; DP2.5; DP3; GS; mAPC; Min; PPP1R46; RATAPC
UniProt ID: (Human) P25054, (Rat) P70478
Entrez Gene ID: (Human) 324, (Mouse) 11789, (Rat) 24205
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.