Novus Biologicals
Manufacturer Code:NBP271585AF350
Catalog # NP2718A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G3 |
Immunogen | Full length recombinant protein of human APBB3 (NP_573419) produced in HEK293T cells. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: amyloid beta (A4) precursor protein-binding family B member 3 amyloid precursor interacting protein Fe65L2 FE65L2amyloid beta A4 precursor protein-binding family B member 3 FE65-like protein 2 MGC150555 MGC87674 Protein Fe65-like 2 SRA; amyloid beta (A4) precursor protein-binding, family B, member 3; amyloid precursor interacting protein; Amyloid-beta A4 precursor protein-binding family B member 3; FE65-like protein 2; Fe65L2; Protein Fe65-like 2
Gene Aliases: APBB3; FE65L2; SRA
UniProt ID: (Human) O95704
Entrez Gene ID: (Human) 10307
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.