Abnova
Manufacturer Code:H00010307B01P
Catalog # 89-002-824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | APBB3 (AAH13158.1 1 a.a. ~ 486 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human APBB3 protein.
For Research Use Only
Protein Aliases: amyloid beta (A4) precursor protein-binding, family B, member 3; amyloid precursor interacting protein; Amyloid-beta A4 precursor protein-binding family B member 3; FE65-like protein 2; FE65L2; MGC150555; MGC87674; Protein Fe65-like 2; SRA
Gene Aliases: APBB3; FE65L2; SRA
UniProt ID: (Human) O95704
Entrez Gene ID: (Human) 10307
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.