Invitrogen
Manufacturer Code:PA521161
Catalog # PIPA521161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide near the center of human AP3S1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 subunit sigma-1; adaptor related protein complex 3, sigma 1 subunit; Adaptor-related protein complex 3 subunit sigma-1; AP-3 complex subunit sigma-1; AP-3 complex subunit sigma-3A; CLAPS3 Sigma3A AP-3 complex sigma-3A subunit AP-3 complex subunit sigma-1 AP-3 complex subunit sigma-3A adapter-related protein complex 3 sigma-1 subunit clathrin-associated/assembly/adapter protein small 3 clathrin-associated/assembly/adaptor protein small 3 (22kD) sigma-3A-adaptin sigma-adaptin 3a sigma3A-adaptin; clathrin adaptor complex AP3, sigma-3A subunit; clathrin-associated/assembly/adapter protein, small 3; Clathrin-associated/assembly/adaptor protein, small 3; clathrin-associated/assembly/adaptor protein, small 3 (22kD); Sigma-3A-adaptin; Sigma-adaptin 3a; Sigma3A-adaptin
Gene Aliases: AP3S1; CLAPS3; Sigma3A
UniProt ID: (Human) Q92572, (Mouse) Q9DCR2
Entrez Gene ID: (Human) 1176, (Mouse) 11777, (Rat) 302290
Molecular Function: membrane traffic protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.