Proteintech
Manufacturer Code:119251AP150UL
Catalog # 50-172-7563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AP3M2 Fusion Protein Ag2572 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 mu-2 subunit; adapter-related protein complex 3 subunit mu-2; adaptor related protein complex 3, mu 2 subunit; Adaptor-related protein complex 3 subunit mu-2; adaptor-related protein complex 3, mu 2 subunit; AP 3 complex subunit mu 2 AP3M2 AP47B CLA20 HA1 47 kDa subunit homolog 2 Mu3B adaptin P47B; AP-3 complex subunit mu-2; clathrin assembly protein assembly protein complex 1 medium chain homolog 2; Clathrin assembly protein assembly protein complex 3 mu-2 medium chain; Clathrin coat assembly protein AP47 homolog 2; Clathrin coat-associated protein AP47 homolog 2; clathrin-associated protein AP47 homolog 2; Golgi adaptor AP-1 47 kDa protein homolog 2; HA1 47 kDa subunit homolog 2; HA1 47kDA subunit homolog 2; m3B; Mu3B-adaptin; P47B
Gene Aliases: 5830445E16Rik; AP-3B; AP3M2; AP47B; CLA20; P47B
UniProt ID: (Human) P53677, (Rat) P53678, (Mouse) Q8R2R9
Entrez Gene ID: (Human) 10947, (Rat) 140667, (Mouse) 64933
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.