Invitrogen
Manufacturer Code:PA521162
Catalog # PIPA521162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human AP3M1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 mu-1 subunit; adapter-related protein complex 3 subunit mu-1; Adaptor-related protein complex 3 subunit mu-1; adaptor-related protein complex AP-3 mu 1 subunit; adaptor-related protein complex AP-3, mu 1 subunit; AP-3 adapter complex mu3A subunit; AP-3 adapter complex mu3A subunit AP-3 complex subunit mu-1 adapter-related protein complex 3 mu-1 subunit clathrin adaptor complex AP3 mu-3A subunit mu-adaptin 3A mu3A-adaptin; AP-3 adaptor complex mu3A subunit; AP-3 complex subunit mu-1; clathrin adaptor complex AP3, mu-3A subunit; clathrin adaptor protein mu3A; clathrin assembly protein assembly protein complex 1 medium chain homolog 1; clathrin assembly protein assembly protein complex 3 mu-1 medium chain; clathrin coat assembly protein AP47 homolog 1; clathrin coat-associated protein AP47 homolog 1; clathrin-associated adaptor protein homolog (p47A); golgi adaptor AP-1 47 kDa protein homolog 1; HA1 47 kDa subunit homolog 1; Mu-adaptin 3A; Mu3A-adaptin
Gene Aliases: 1200013D09Rik; AP3M1; C78982; P47a; R75378
UniProt ID: (Human) Q9Y2T2, (Mouse) Q9JKC8
Entrez Gene ID: (Human) 26985, (Rat) 171126, (Mouse) 55946
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.