Invitrogen
Manufacturer Code:PA5100384
Catalog # PIPA5100384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AP3D1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA407035 adapter-related protein complex 3 subunit delta-1 adaptin delta adaptor related protein complex 3 delta 1 subunit adaptor related protein complex 3 delta 1 subunit adaptor-related protein complex 3 subunit delta-1 adaptor-related protein complex 3 delta 1 subunit adaptor-related protein complex 3 delta subunit adaptor-related protein complex AP-3 delta subunit ADTD ap3 AP-3 complex delta subunit partial CDS AP-3 complex subunit delta AP-3 complex subunit delta-1 Ap3d Ap3d1 Bolvr delta adaptin delta-adaptin delta-adaptin partial CDS hBLVR mBLVR1 mh mocha PRO0039 subunit of putative vesicle coat adaptor complex AP-3; adapter-related protein complex 3 subunit delta-1; adaptin, delta; adaptor related protein complex 3, delta 1 subunit; Adaptor-related protein complex 3 subunit delta-1; adaptor-related protein complex 3, delta 1 subunit; adaptor-related protein complex 3, delta subunit; adaptor-related protein complex AP-3, delta subunit; AP-3 complex delta subunit, partial CDS; AP-3 complex subunit delta; AP-3 complex subunit delta-1; delta adaptin; Delta-adaptin; delta-adaptin, partial CDS; mBLVR1; subunit of putative vesicle coat adaptor complex AP-3
Gene Aliases: AA407035; ADTD; Ap3d; AP3D1; Bolvr; hBLVR; mBLVR1; mh; mocha; PRO0039
UniProt ID: (Human) O14617, (Mouse) O54774
Entrez Gene ID: (Human) 8943, (Mouse) 11776, (Rat) 314633
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.