Novus Biologicals
Manufacturer Code:NBP29204620UL
Catalog # NB025985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 520-700 of human AP3D1 (NP_003929.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 subunit delta-1; Adapter-related protein complex 3 subunit delta-1 adaptin delta adaptor-related protein complex 3 delta 1 subunit ADTD AP-3 complex delta subunit partial CDS AP-3 complex subunit delta AP-3 complex subunit delta-1 delta adaptin Delta-adaptin delta-adaptin partial CDS hBLVR subunit of putative vesicle coat adaptor complex AP-3; adaptin, delta; adaptor related protein complex 3, delta 1 subunit; Adaptor-related protein complex 3 subunit delta-1; adaptor-related protein complex 3, delta 1 subunit; AP-3 complex delta subunit, partial CDS; AP-3 complex subunit delta; AP-3 complex subunit delta-1; delta adaptin; Delta-adaptin; delta-adaptin, partial CDS; subunit of putative vesicle coat adaptor complex AP-3
Gene Aliases: ADTD; AP3D1; hBLVR; PRO0039
UniProt ID: (Human) O14617
Entrez Gene ID: (Human) 8943
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.