Novus Biologicals
Manufacturer Code:NBP176590
Catalog # NBP176590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human AP3B2. The immunogen is located within the first 50 amino acids of AP3B2. Amino Acid Squence: IFSSDYKRHDDLKEMLDT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 subunit beta-2; Adapter-related protein complex 3 subunit beta-2 Adaptor protein complex AP-3 subunit beta-2 adaptor-related protein complex 3 beta 2 subunit beta-3B-adaptin Clathrin assembly protein complex 3 beta-2 large chain DKFZp686D17136 NAPTBAP-3 complex subunit beta-2 Neuronal adaptin-like protein beta-subunit Neuron-specific vesicle coat protein beta-NAP; Adaptor protein complex AP-3 subunit beta-2; adaptor related protein complex 3, beta 2 subunit; Adaptor-related protein complex 3 subunit beta-2; adaptor-related protein complex 3, beta 2 subunit; AP-3 complex subunit beta-2; Beta-3B-adaptin; Clathrin assembly protein complex 3 beta-2 large chain; Neuron-specific vesicle coat protein beta-NAP; Neuronal adaptin-like protein, beta-subunit
Gene Aliases: AP3B2; NAPTB
UniProt ID: (Human) Q13367
Entrez Gene ID: (Human) 8120
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.