Proteintech
Manufacturer Code:183311AP150UL
Catalog # 50-173-2118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AP3B2 Fusion Protein Ag12861 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 subunit beta-2; adaptor protein complex AP-3 beta-2 subunit; Adaptor protein complex AP-3 subunit beta-2; adaptor related protein complex 3, beta 2 subunit; Adaptor-related protein complex 3 subunit beta-2; adaptor-related protein complex 3, beta 2 subunit; adaptor-related protein complex AP-3, beta 2 subunit; AP 3 complex subunit beta 2 AP3B2 Beta 3B adaptin NAPTB; AP-3 complex beta-2 subunit; AP-3 complex subunit beta-2; Beta-3B-adaptin; beta3B-adaptin; Clathrin assembly protein complex 3 beta-2 large chain; Neuron-specific vesicle coat protein beta-NAP; Neuronal adaptin-like protein, beta-subunit
Gene Aliases: [b]-NAP; AI549966; AP3B2; AU042881; beta-NAP; beta3B; NAPTB
UniProt ID: (Human) Q13367, (Mouse) Q9JME5
Entrez Gene ID: (Human) 8120, (Mouse) 11775, (Rat) 308777
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.