Invitrogen
Manufacturer Code:PA576207
Catalog # PIPA576207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human AP3B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; Adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3, beta 1 subunit; Adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; Beta-3A-adaptin; beta3A-adaptin; Clathrin assembly protein complex 3 beta-1 large chain; PE HPS HPS2 ADTB3 ADTB3A; recombination induced mutation 2
Gene Aliases: ADTB3; ADTB3A; AP-3; AP3B1; AU015684; beta3A; C78395; HPS; HPS2; PE; pearl; rim2
UniProt ID: (Human) O00203, (Mouse) Q9Z1T1
Entrez Gene ID: (Human) 8546, (Mouse) 11774
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.