Invitrogen
Manufacturer Code:PA5106298
Catalog # PIPA5106298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AP3B1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 3 subunit beta-1; adapter-related protein complex 3 subunit beta-1 adaptor protein complex AP-3 beta-1 subunit adaptor protein complex AP-3 subunit beta-1 adaptor related protein complex 3 beta 1 subunit adaptor related protein complex 3 beta 1 subunit adaptor-related protein complex 3 subunit beta-1 adaptor-related protein complex 3 beta 1 subunit adaptor-related protein complex AP-3 beta 1 subunit ADTB3 ADTB3A ap3 AP-3 AP-3 complex beta-3A subunit AP-3 complex subunit beta-1 AP3B1 AU015684 beta3A Beta-3A-adaptin beta3A-adaptin C78395 Clathrin assembly protein complex 3 beta-1 large chain HPS Hps2 pe pearl recombination induced mutation 2 rim2; adaptor protein complex AP-3 beta-1 subunit; Adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3, beta 1 subunit; Adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; Beta-3A-adaptin; beta3A-adaptin; Clathrin assembly protein complex 3 beta-1 large chain; recombination induced mutation 2
Gene Aliases: ADTB3; ADTB3A; AP-3; AP3B1; AU015684; beta3A; C78395; HPS; HPS2; PE; pearl; rim2
UniProt ID: (Human) O00203, (Mouse) Q9Z1T1
Entrez Gene ID: (Human) 8546, (Mouse) 11774, (Rat) 309969
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.