Invitrogen
Manufacturer Code:PA520745
Catalog # PIPA520745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the carboxy terminus of human AP2M1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 2 mu subunit; adapter-related protein complex 2 subunit mu; Adaptin-mu2; Adaptor protein complex AP-2 subunit mu; adaptor related protein complex 2, mu 1 subunit; Adaptor-related protein complex 2 subunit mu; adaptor-related protein complex 2, mu 1 subunit; AP-2 complex subunit mu; AP-2 mu 2 chain; AP-2 mu chain; AP50 CLAPM1 mu2 AP-2 complex subunit mu AP-2 mu 2 chain AP-2 mu chain HA2 50 kDA subunit adapter-related protein complex 2 mu subunit adaptin-mu2 adaptor protein complex AP-2 subunit mu clathrin adaptor complex AP2 mu subunit clathrin assembly protein complex 2 medium chain clathrin coat adaptor protein AP50 clathrin coat assembly protein AP50 clathrin coat-associated protein AP50 clathrin-associated/assembly/adaptor protein medium 1 plasma membrane adaptor AP-2 50 kDa protein; clathrin adaptor complex AP2, mu subunit; clathrin assembly protein complex 2 medium chain; Clathrin assembly protein complex 2 mu medium chain; clathrin coat adaptor protein AP50; Clathrin coat assembly protein AP50; Clathrin coat-associated protein AP50; clathrin-associated/assembly/adaptor protein, medium 1; coat assembly protein complex 50 kD; HA2 50 kDA subunit; Mu2-adaptin; Plasma membrane adaptor AP-2 50 kDa protein; plasma membrane adaptor AP-2 50kDA protein
Gene Aliases: AP2M1; AP50; CLAPM1; KIAA0109; mu2
UniProt ID: (Human) Q96CW1, (Rat) P84092, (Mouse) P84091
Entrez Gene ID: (Human) 1173, (Rat) 116563, (Mouse) 11773
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.