Abnova
Manufacturer Code:H00000163M01
Catalog # 89-016-061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D5 |
Immunogen | AP2B1 (NP_001273.1 585 a.a. ~ 651 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant AP2B1.
For Research Use Only
Protein Aliases: adapter-related protein complex 2 beta subunit; adapter-related protein complex 2 subunit beta; adaptin, beta 2 (beta); Adaptor protein complex AP-2 subunit beta; adaptor related protein complex 2, beta 1 subunit; Adaptor-related protein complex 2 subunit beta; adaptor-related protein complex 2, beta 1 subunit; ADTB2; AP-2 complex subunit beta; AP105B; AP2-BETA; Beta-2-adaptin; Beta-adaptin; CLAPB1; Clathrin assembly protein complex 2 beta large chain; clathrin-associated/assembly/adaptor protein, large, beta 1; DKFZp781K0743; Plasma membrane adaptor HA2/AP2 adaptin beta subunit; testicular tissue protein Li 22
Gene Aliases: ADTB2; AP105B; AP2-BETA; AP2B1; CLAPB1
UniProt ID: (Human) P63010
Entrez Gene ID: (Human) 163
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.