Invitrogen
Manufacturer Code:MA190265
Catalog # PIMA190265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A6/2/2 |
Immunogen | C-terminal peptide of AP-2 alpha (PNSHTDNNAKSSDKEEKHRK). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta; Activating enhancer-binding protein 2-alpha; Activating enhancer-binding protein 2-beta; Activator protein 2; AP-2; AP-2 AP-2alpha AP2TF BOFS FLJ51761 TFAP2; AP-2 transcription factor; AP2-alpha; AP2-beta; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2 beta (activating enhancer binding protein 2 beta); Transcription factor AP-2-alpha; Transcription factor AP-2-beta
Gene Aliases: AP-2; AP-2alpha; AP-2B; AP2-B; AP2TF; BOFS; TFAP2; TFAP2A; TFAP2B
UniProt ID: (Human) P05549, (Human) Q92481
Entrez Gene ID: (Human) 7020, (Human) 7021
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.