Novus Biologicals
Manufacturer Code:NB1002639AF647
Catalog # NB1002639AH
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AP2a 8G8/5 |
Immunogen | Bacterialy expressed AP-2a/AP-2g fusion protein. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activating enhancer-binding protein 2-alpha; Activating enhancer-binding protein 2-alpha Activator protein 2 AP-2 transcription factor AP2-alpha AP2TFAP-2alpha AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha) BOFS FLJ51761 TFAP2 transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha) transcription factor AP-2-alpha; Activator protein 2; AP-2; AP-2 transcription factor; AP2-alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha
Gene Aliases: AP-2; AP-2alpha; AP2TF; BOFS; TFAP2; TFAP2A
UniProt ID: (Human) P05549
Entrez Gene ID: (Human) 7020
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.