Invitrogen
Manufacturer Code:MA1872
Catalog # 01-670-128
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B5 |
Immunogen | N-terminus of human AP2a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer-binding protein 2 alpha; Activating enhancer-binding protein 2-alpha; Activator protein 2; AP-2; AP-2 transcription factor; AP2-alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha; transcription factor AP2
Gene Aliases: AP-2; Ap-2 (a); AP-2alpha; Ap2; AP2alpha; AP2TF; BOFS; Tcfap2a; TFAP2; TFAP2A
UniProt ID: (Human) P05549, (Mouse) P34056
Entrez Gene ID: (Human) 7020, (Chicken) 395982, (Mouse) 21418
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.