Invitrogen
Manufacturer Code:PA599259
Catalog # PIPA599259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AP-2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2 alpha Activating enhancer-binding protein 2-alpha Activator protein 2 Ap2 AP-2 Ap-2 (a) AP-2 transcription factor AP2alpha AP-2alpha AP2-alpha Ap2tf BOFS ERF-1 FLJ51761 OTTHUMP00000214240 OTTHUMP00000214243 Tcfap2a TFAP2 TFAP2A transcription factor AP-2 alpha transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha) transcription factor AP-2 alpha transcription factor AP-2-alpha transcription factor AP-2-alpha-like; Activating enhancer-binding protein 2-alpha; Activator protein 2; AP-2; AP-2 transcription factor; AP2-alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha
Gene Aliases: AP-2; Ap-2 (a); AP-2alpha; Ap2; AP2alpha; AP2TF; BOFS; Tcfap2a; TFAP2; TFAP2A
UniProt ID: (Human) P05549, (Mouse) P34056
Entrez Gene ID: (Human) 7020, (Mouse) 21418, (Rat) 306862
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.