Invitrogen
Manufacturer Code:PA529769
Catalog # PIPA529769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 236 and 437 of Human AP2 alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating enhancer-binding protein 2-alpha; Activator protein 2; AP-2; AP-2 AP-2alpha AP2TF BOFS TFAP2 AP-2 transcription factor AP2-alpha activating enhancer-binding protein 2-alpha activator protein 2 transcription factor AP-2-alpha; AP-2 transcription factor; AP2-alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha
Gene Aliases: AP-2; AP-2alpha; AP2TF; BOFS; TFAP2; TFAP2A
UniProt ID: (Human) P05549
Entrez Gene ID: (Human) 7020
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.