Invitrogen
Manufacturer Code:PA125327
Catalog # PIPA125327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH corresponding to residues S(420) H T D N N A K S S D K E E K H R K(437) of human AP2 alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer-binding protein 2 alpha; Activating enhancer-binding protein 2-alpha; Activator protein 2; AP-2; AP-2 AP-2alpha AP2TF BOFS FLJ51761 TFAP2; AP-2 transcription factor; AP2-alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha; transcription factor; AP-2
Gene Aliases: AP-2; Ap-2 (a); AP-2alpha; AP2; AP2alpha; AP2TF; BOFS; Tcfap2a; TFAP2; TFAP2A
UniProt ID: (Sheep) Q9N0N3, (Human) P05549, (Mouse) P34056
Entrez Gene ID: (Sheep) 443109, (Human) 7020, (Mouse) 21418, (Rat) 306862
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.