Proteintech
Manufacturer Code:169321AP150UL
Catalog # 50-173-1270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AP1B1 Fusion Protein Ag10417 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adapter-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1; adaptor protein complex AP-1, beta 1 subunit; adaptor-related protein complex 1 beta 1 subunit; Adaptor-related protein complex 1 subunit beta-1; adaptor-related protein complex 1, beta 1 subunit; adaptor-related protein complex AP-1 beta 1 subunit; adaptor-related protein complex AP-1, beta 1 subunit; ADTB1 AP 1 complex subunit beta 1 AP105A AP1B1 BAM22 Beta 1 adaptin Beta adaptin 1 CLAPB2; AP-1 complex subunit beta-1; Beta-1-adaptin; Beta-adaptin 1; beta-prime adaptin; beta-prime-adaptin; beta1-adaptin; Clathrin assembly protein complex 1 beta large chain; Golgi adaptor HA1/AP1 adaptin beta subunit; Mutant line 1660; plasma membrane adaptor HA2/AP2 adaptor beta subunit
Gene Aliases: ADTB1; AP105A; AP1B1; b2b1660Clo; BAM22; CLAPB2
UniProt ID: (Human) Q10567, (Rat) P52303, (Mouse) O35643
Entrez Gene ID: (Human) 162, (Rat) 29663, (Mouse) 11764
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.