Novus Biologicals
Manufacturer Code:NB1002640C
Catalog # NB1002640C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AP2b beta 1F3/8 |
Immunogen | Truncated AP-2Beta protein prepared from bacteria |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta; activating enhancer binding protein 2 beta Activating enhancer-binding protein 2-beta AP-2B AP2-B AP2-beta CHAR MGC21381 transcription factor AP-2 beta (activating enhancer binding protein 2 beta) transcription factor AP-2 beta (activating enhancer-binding protein 2 beta) transcription factor AP-2-beta; Activating enhancer-binding protein 2-beta; AP2-beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); Transcription factor AP-2-beta
Gene Aliases: AP-2B; AP2-B; TFAP2B
UniProt ID: (Human) Q92481
Entrez Gene ID: (Human) 7021
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.