R&D Systems
Manufacturer Code:AF5634SP
Catalog # AF5634SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human AP-2 beta Leu121-Phe218 Accession # Q92481 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta; activating enhancer binding protein 2 beta Activating enhancer-binding protein 2-beta AP2 beta AP-2B AP2-B AP2-beta CHAR MGC21381 TFAP2B transcription factor AP-2 beta (activating enhancer binding protein 2 beta) transcription factor AP-2 beta (activating enhancer-binding protein 2 beta) transcription factor AP-2-beta; Activating enhancer-binding protein 2-beta; AP2-beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); Transcription factor AP-2-beta
Gene Aliases: AP-2B; AP2-B; TFAP2B
UniProt ID: (Human) Q92481
Entrez Gene ID: (Human) 7021
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.