Invitrogen
Manufacturer Code:PA5101413
Catalog # PIPA5101413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AOX1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI196512 AI255253 Aldehyde oxidase aldehyde oxidase (female form) Aldehyde oxidase 1 aldehyde oxidase 2 Ao AOH1 AOX1 Aox-1 Aox2 Aox-2 azaheterocycle hydroxylase Azaheterocycle hydroxylase 1 Moro Retinal oxidase Ro; Aldehyde oxidase; aldehyde oxidase (female form); Aldehyde oxidase 1; aldehyde oxidase 2; Azaheterocycle hydroxylase; Azaheterocycle hydroxylase 1; Retinal oxidase
Gene Aliases: AI196512; AI255253; AO; AOH1; Aox-1; Aox-2; AOX1; Aox2; Moro; Ro
UniProt ID: (Human) Q06278, (Mouse) O54754, (Rat) Q9Z0U5
Entrez Gene ID: (Human) 316, (Mouse) 11761, (Rat) 54349
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.