Novus Biologicals
Manufacturer Code:NBP281895
Catalog # NB017206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AOF1 antibody was raised against a 17 amino acid peptide near the amino terminus of human AOF1. The immunogen is located within amino acids 80 - 130 of AOF1. Amino Acid Squence: FDHYYRSHKDGYDKYT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amine oxidase (flavin containing) domain 1; amine oxidase, flavin containing 1; AOF1 bA204B7.3 C6orf193 dJ298J15.2 DKFZp686I0412 flavin containing 1 flavin-containing amine oxidase domain-containing protein 1 FLJ33898 FLJ34109 FLJ43328 LSD2 lysine (K)-specific demethylase 1B lysine-specific histone demethylase 1B lysine-specific histone demethylase 2; Flavin-containing amine oxidase domain-containing protein 1; lysine (K)-specific demethylase 1B; Lysine-specific histone demethylase 1B; Lysine-specific histone demethylase 2
Gene Aliases: AOF1; bA204B7.3; C6orf193; dJ298J15.2; KDM1B; LSD2
UniProt ID: (Human) Q8NB78
Entrez Gene ID: (Human) 221656
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.