Abnova
Manufacturer Code:PAB22537
Catalog # 89-124-246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human AOF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant AOF1.
For Research Use Only
Protein Aliases: amine oxidase (flavin containing) domain 1; amine oxidase, flavin containing 1; bA204B7.3; C6orf193; dJ298J15.2; DKFZp686I0412; Flavin-containing amine oxidase domain-containing protein 1; FLJ33898; FLJ34109; FLJ43328; lysine (K)-specific demethylase 1B; Lysine-specific histone demethylase 1B; Lysine-specific histone demethylase 2
Gene Aliases: AOF1; bA204B7.3; C6orf193; dJ298J15.2; KDM1B; LSD2
UniProt ID: (Human) Q8NB78
Entrez Gene ID: (Human) 221656
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.