Invitrogen
Manufacturer Code:PA569361
Catalog # PIPA569361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human AOC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amine oxidase [copper-containing]; amine oxidase copper containing 2; amine oxidase, copper containing 2 (retina-specific); RAO; RAO amine oxidase copper containing 2 retina-specific DAO2; retina-specific amine oxidase; Retina-specific copper amine oxidase; Semicarbazide-sensitive amine oxidase; SSAO
Gene Aliases: AOC2; DAO2; RAO
UniProt ID: (Human) O75106, (Mouse) Q812C9
Entrez Gene ID: (Bovine) 100336082, (Horse) 100630218, (Dog) 490957, (Human) 314, (Pig) 100520513, (Mouse) 237940
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.