Abnova
Manufacturer Code:H00000314K
Catalog # 89-200-275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AOC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human AOC2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Amine oxidase [copper-containing]; amine oxidase copper containing 2; amine oxidase, copper containing 2 (retina-specific); DAO2 RAO; RAO; Retina-specific copper amine oxidase; Semicarbazide-sensitive amine oxidase; SSAO
Gene Aliases: AOC2; DAO2; RAO
UniProt ID: (Human) O75106
Entrez Gene ID: (Human) 314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.