Proteintech
Manufacturer Code:154161AP150UL
Catalog # 50-173-0211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANXA9 Fusion Protein Ag7702 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: annexin 31; Annexin 31 Annexin 9 annexin A9 Annexin XXXI ANX31 ANXA9 Pemphaxin; Annexin A9; Annexin XXXI; Annexin-31; Annexin-9; Pemphaxin
Gene Aliases: 1110003P15Rik; 2310069F17Rik; ANX31; ANXA9
UniProt ID: (Human) O76027, (Mouse) Q9JHQ0
Entrez Gene ID: (Human) 8416, (Mouse) 71790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.