Proteintech
Manufacturer Code:167231AP150UL
Catalog # 50-173-1139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANTXR2 Fusion Protein Ag10162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anthrax toxin receptor 2; anthrax toxin receptor 2 ANTXR2 CMG 2 CMG2 ISH JHF; Capillary morphogenesis gene 2 protein; capillary morphogenesis protein 2; CMG-2
Gene Aliases: 2310046B19Rik; ANTXR2; AW561899; cI-35; CMG-2; CMG2; HFS; ISH; JHF
UniProt ID: (Human) P58335, (Mouse) Q6DFX2
Entrez Gene ID: (Human) 118429, (Rat) 305633, (Mouse) 71914
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.