Proteintech
Manufacturer Code:150911AP150UL
Catalog # 50-172-9971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ANTXR1 Fusion Protein Ag4626 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310008J16Rik; 2810405N18Rik; Anthrax toxin receptor 1; anthrax toxin receptor 1 ANTXR1 ATR TEM8 Tumor endothelial marker 8; Tumor endothelial marker 8
Gene Aliases: 2310008J16Rik; 2810405N18Rik; Antrx1; ANTXR1; ATR; GAPO; TEM8
UniProt ID: (Human) Q9H6X2, (Rat) Q0PMD2, (Mouse) Q9CZ52
Entrez Gene ID: (Human) 84168, (Rat) 362393, (Mouse) 69538
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.