Invitrogen
Manufacturer Code:PA5109394
Catalog # PIPA5109394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human ANT1 (NP_001142.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700034J06Rik 2F1 4930433D19Rik 4933433F13Rik AAC1 AAC2 AAC4 Adenine nucleotide translocator 1 Adenine nucleotide translocator 2 Adenine nucleotide translocator 4 ADPATP carrier protein 1 ADPATP carrier protein 2 ADPATP carrier protein 4 ADPATP carrier protein fibroblast isoform ADPATP carrier protein heart/skeletal muscle isoform T1 ADP/ATP translocase 1 ADP/ATP translocase 2 ADP/ATP translocase 2 N-terminally processed ADP/ATP translocase 4 Anc1 ANT ANT 1 ANT 2 ANT 4 ANT1 ANT2 Ant4 AU019225 AW557176 COXPD28 D6Bwg0781e mANC1 Mitochondrial S-adenosylmethionine transporter MTDPS12 PEO2 PEO3 PEOA2 S-adenosylmethionine mitochondrial carrier protein Samc SFEC SFEC35kDa SLC25A26 Slc25a31 SLC25A4 SLC25A5 Slc25a6 Solute carrier family 25 member 26 Solute carrier family 25 member 31 Solute carrier family 25 member 4 Solute carrier family 25 member 5 Sperm flagellar energy carrier protein SZGENESYMBOL2 T1 T2 T3; Adenine nucleotid translocator 2 fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotid translocator 2, fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocase 4; adenine nucleotide translocase-1; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); adenine nucleotide translocator 2 fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 2, fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 1; ADP,ATP carrier protein 2; ADP,ATP carrier protein 4; ADP,ATP carrier protein, fibroblast isoform; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP translocase 1; ADP/ATP translocase 2; ADP/ATP translocase 2, N-terminally processed; ADP/ATP translocase 4; ANT 1; ANT 2; ANT 4; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; Mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; Solute carrier family 25 member 26; Solute carrier family 25 member 31; Solute carrier family 25 member 4; Solute carrier family 25 member 5; solute carrier family 25, member 26; Sperm flagellar energy carrier protein; SZFULLNAME2
Gene Aliases: 1700034J06Rik; 2F1; 4930433D19Rik; 4933433F13Rik; AAC1; AAC2; AAC4; Anc1; ANT; ANT 1; ANT 4; ANT1; ANT2; ANT4; AU019225; AW557176; COXPD28; D6Bwg0781e; MTDPS12; PEO2; PEO3; PEOA2; SAMC; SFEC; SFEC35kDa; SLC25A26; SLC25A31; SLC25A4; SLC25A5; Slc25a6; SZGENESYMBOL2; T1; T2; T3
UniProt ID: (Human) Q70HW3, (Human) Q9H0C2, (Human) P12235, (Human) P05141, (Mouse) Q5U680, (Mouse) Q3V132, (Rat) Q05962, (Mouse) P48962, (Rat) Q09073, (Mouse) P51881
Entrez Gene ID: (Human) 115286, (Human) 83447, (Human) 291, (Human) 292, (Mouse) 67582, (Rat) 362403, (Rat) 689108, (Mouse) 73333, (Rat) 85333, (Mouse) 11739, (Rat) 25176, (Mouse) 11740
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.